missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 300 to 30 C4

Product Code. 10505951 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
0.1 kg
1 kg
Unit Size:
1000g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10505951 1 kg 1000g
10276211 0.1 kg 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10505951 Supplier Macherey-Nagel™ Supplier No. 711690.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Particle Size 25 to 40 μm
Pore Size 300 Å
Stationary Phase POLYGOPREP 300 to 30
pH 2 to 8
Quantity 1 kg
Sorbent POLYGOPREP C4
Type Polygoprep
For Use With (Application) Liquid Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.