Learn More
Macherey-Nagel™ Polygoprep™ 60 to 30 NH2
Product Code. 10388051 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
0.1 kg
1 kg
Unit Size:
1000g
100g
Product Code. 10388051
Brand: Macherey-Nagel™ 711013.1000
Specifications
Particle Size 15 to 25 μm
Pore Size 60 Å
Stationary Phase POLYGOPREP 60 to 20
Technique Liquid Chromatography
pH 2 to 8
Quantity 1 kg
Sorbent POLYGOPREP NH2
Surface Area (Metric) 350 m2/g
Type Polygoprep
Material Silica
For Use With (Application) Liquid Chromatography