Learn More
Macherey-Nagel™ Polygram POLYAMIDE-6 Polyester Sheets
Brand:  Macherey-Nagel™ 803013
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10496661
  • € 221.00 / Pack of 25
Estimated Shipment: 24-05-2024
to see stock.
Specifications
25 Pk.
Polyester Sheet
0.1 mm
20 x 20 cm