missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polymaleic Acid (Aqueous Solution), TRC
Specifications
Specifications
| Chemical Name or Material | Polymaleic Acid |
| CAS | 26099-09-2 |
| Molecular Formula | (C4H4O4)n |
| Quantity | 2.5 g |
| Synonym | 2-Butenedioic acid (Z)-','homopolymer (ZCI), Maleic acid','polymers (8CI), Accent T 1107, Acumer 4200, Aron A 6510, Bel 200 premix, Belclene 200, Belclene 200LA, Belclene 710, Dequest P 9000, DP 3328, Maleic acid homopolymer, Maleic acid polymer, Nonpol PWA 50W, Poly(maleic acid), SH 150 |
| InChI Formula | InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1- |
| SMILES | O=C(O)/C([H])=C([H])\C(O)=O |
| IUPAC Name | Poly(maleic acid) |
| Molecular Weight (g/mol) | 146.141 |
| Formula Weight | 146.058 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?