missing translation for 'onlineSavingsMsg'
Learn More

Polymaleic Acid (Aqueous Solution), TRC

Product Code. 30418518
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418518 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418518 Supplier Toronto Research Chemicals Supplier No. TRCP6894081G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polymaleic Acid
CAS 26099-09-2
Molecular Formula (C4H4O4)n
Quantity 1 g
Synonym 2-Butenedioic acid (Z)-','homopolymer (ZCI), Maleic acid','polymers (8CI), Accent T 1107, Acumer 4200, Aron A 6510, Bel 200 premix, Belclene 200, Belclene 200LA, Belclene 710, Dequest P 9000, DP 3328, Maleic acid homopolymer, Maleic acid polymer, Nonpol PWA 50W, Poly(maleic acid), SH 150
InChI Formula InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-
SMILES O=C(O)/C([H])=C([H])\C(O)=O
IUPAC Name Poly(maleic acid)
Molecular Weight (g/mol) 146.141
Formula Weight 146.058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.