Learn More
Vitlab™ Polymethylpentene Forceps
Manufactured with PMP for very good resilience. VITLab™ Polymethylpentene Forceps are ideal for use in bottling and decanting.
Brand: Vitlab™ 67895
19.16 EUR valid until 2024-12-31
Use promo code "23277" to get your promotional price.
Additional Details : Weight : 0.03060kg
Description
- White, Pointed, Elastic
Specifications
Pointed | |
115 mm | |
10/Pack | |
Forceps |
PMP | |
Forceps | |
For autoclaving and microwaves, bottling and decanting |