missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polynuclear Aromatic Hydrocarbons Antibody (BAP-13), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-479B
This item is not returnable.
View return policy
Description
Polynuclear Aromatic Hydrocarbons Monoclonal antibody specifically detects Polynuclear Aromatic Hydrocarbons in Nonspecies specific samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Polynuclear Aromatic Hydrocarbons | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Multi-step | |
| RUO | |
| Non-species specific | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| BAP-13 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| PAHs | |
| Benzo[a]pyrenyl-1-butyric acid conjugated to BSA. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction