missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyoma Virus Early Antigen Antibody (PyC), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-2793PCP
This item is not returnable.
View return policy
Description
Polyoma Virus Early Antigen Monoclonal antibody specifically detects Polyoma Virus Early Antigen in Virus samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Polyoma Virus Early Antigen | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Immunocytochemistry, Immunoprecipitation | |
| PyC | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Large T antigen, Middle T antigen, Polyoma virus large T antigen, Polyoma virus middle T antigen, Polyoma virus small T antigen, Polyomavirus early antigen, Polyomavirus large T antigen, Polyomavirus middle t antigen, Polyomavirus small T antigen, Small T antigen | |
| Polyoma virus-transformed Wistar rat fibroblast cellline REWA5/T1A1. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction