missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyoma Virus, Large T Antigen Antibody (PyLT), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-2755AF488
This item is not returnable.
View return policy
Description
Polyoma Virus, Large T Antigen Monoclonal specifically detects Polyoma Virus, Large T Antigen in Virus samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Polyoma Virus, Large T Antigen | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| PyLT | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Polyoma virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Polyoma virus, Large T antigen (PyLT). | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction