Learn More
Kartell™ Polyoxymethylene Clamps
Manufactured from polyoxymethylene for low friction and excellent dimensional stability. Kartell™ Polyoxymethylene Clamps are color coded according to joint size and provide a safe connection for glass conical joints.
Brand: Kartell™ 3192600
Additional Details : Weight : 0.03400kg
Description
- Plastic construction generates a corrosion proof device that does not scratch glass
- Available in various colors that correspond with the joint size of the connection
- Autoclavable
- Safe connecction for standard-finish glass conical joints
- Plastic construction generates a corrosion proof device that does not scratch glass
- Available in various colors that correspond with the joint size of the connection
- Autoclavable
- Safe connecction for standard-finish glass conical joints
Specifications
1/Pk. | |
POM | |
Autoclavable |
Joint Clamp | |
Standard-finish glass conical joint size 19/26 | |
Light Blue |