missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polypeptide GalNac Transferase 7/GALNT7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | Polypeptide GalNac Transferase 7/GALNT7 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617395
|
Novus Biologicals
NBP2-39054-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18145569
|
Novus Biologicals
NBP2-39054 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Polypeptide GalNac Transferase 7/GALNT7 Polyclonal specifically detects Polypeptide GalNac Transferase 7/GALNT7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Polypeptide GalNac Transferase 7/GALNT7 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q86SF2 | |
| 51809 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DDPSPLSRMREDRDVNDPMPNRGGNGLAPGEDRFKPVVPWPHVEGVEVDLESIRRINKAKNEQEHHAGG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.4.1, EC 2.4.1.-, EC 2.4.1.37, EC 2.4.1.41, GalNAcT7, GalNAc-T7, N-acetylgalactosaminyltransferase 7, Polypeptide GalNAc transferase 7, polypeptide N-acetylgalactosaminyltransferase 7, pp-GaNTase 7, Protein-UDP acetylgalactosaminyltransferase 7, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 7, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 7 (GalNAc-T7) | |
| GALNT7 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title