Learn More
BRAND™ Urine Beaker, PP, with Screw Cap PE, CE-IVD
Shop All Brand Products
Click to view available options
Closure Color:
Green
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 240
Description
- PP, screw cap PE.
- For hygienic urine sample handling.
- CE marked according to guideline EU IVDR 2017/746
Specifications
Specifications
| Product Type | Urine Beaker |
| Closure Color | Green |
| Closure Type | Screw Cap |
| Graduated | Yes |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Material | Polyethylene |
| Color | Transparent |
| For Use With (Application) | Hygienic urine sample handling |
| Height (Metric) | 75 mm |
| Show More |