Learn More
Cole-Parmer™ Polypropylene 3-Way Two-Position Elliptic Valve
Brand: Cole-Parmer™ 98150-00
Additional Details : Weight : 0.80000kg
Specifications
3-Way Elliptic Valve | |
For 1/8 in. NPT(F) connectors | |
1 Pack |
Polypropylene | |
For liquid, gas, and vacuum applications |