Learn More
Eppendorf™ Polypropylene Bar-coded 384 Well DNA LoBind Microplate
Brand: Eppendorf™ 0030610326
Additional Details : Weight : 3.51000kg
Specifications
16 x 24 | |
Polypropylene | |
80 |
Clear | |
384 | |
80/Pk. |