Learn More
Bel-Art™ SP Scienceware™ Polypropylene Basket
Shop All SP Scienceware Products
Click to view available options
Dimensions (L x W x H):
229 x 229 x 229 mm
Unit Size:
Each
Description
- Polypropylene construction
- For use with rinsing, storage and transport of test tubes and other laboratory glassware
- Steam autoclavable at 121°C (250°F)
Specifications
Specifications
| Product Type | Basket |
| Material | Polypropylene |
| Quantity | 1 Each |
| Size | 229 x 229 x 229 mm |
| Dimensions (L x W x H) | 229 x 229 x 229 mm |