Learn More
VITLAB™ Polypropylene Bowls
Product Code. 10693971
Click to view available options
Capacity (Metric):
3 L
9 L
13 L
Includes:
3 Bowls
5 Bowls
Unit Size:
Each
Pack of 3
Pack of 5
Product Code. 10693971
Brand: VITLAB™ 43094
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mix, store and save lab and foodstuff products. VITLAB™ Polypropylene Bowls are sturdy, lightweight bowls designed for stability and durability. Constructed of high-quality white polypropylene in a variety of sizes. Suitable for use with foodstuffs.
Mix, store and save lab and foodstuff products. VITLAB™ Polypropylene Bowls are sturdy, lightweight bowls designed for stability and durability. Constructed of high-quality white polypropylene in a variety of sizes. Suitable for use with foodstuffs.
Specifications
Bottom Shape Flat
Capacity (Metric) 9 L
Color White
Material Polypropylene
Shape Round
Type Bowl Set
Includes 3 Bowls
Height (Metric) 150 mm
Diameter (Metric) 360 mm