Learn More
VITLAB™ Polypropylene Bucket with Spout
Click to view available options
Unit Size:
Each
Description
- Transparent
- Divided into 1 liter segments
- Stable handle and spout for easy emptying
- Highly resistant to chemicals
- Without lid
Specifications
Specifications
| Product Type | Graduated Bucket |
| Material | Polypropylene |
| Color | Transparent |
| Diameter (Metric) Outer | 310 mm |
| Height (Metric) | 330 mm |