Learn More
Wesbart™ Polypropylene Centrifuge Tube Box
Product Code. 15971231
Change view
Click to view available options
Holds:
25 (15 mL Falcon Tubes)
9 (50 mL Falcon Tubes)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15971231 25 (15 mL Falcon Tubes) Each
15981231 9 (50 mL Falcon Tubes) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15971231 Supplier Wesbart™ Supplier No. PA128/2515
For use in ULT freezers down to a temperature of -86 degrees
  • Polypropylene
  • Holds Falcon centrifuge tubes
  • All boxes have an external footprint of 133 x 133mm
  • Plastic dividers are also available to help organize your samples
  • All boxes are 128mm high x 133mm wide x 133mm deep
Specifications
Type Cryo Storage Box
For Use With (Equipment) Centrifuge Tubes
Holds 25 (15 mL Falcon Tubes)
Length (Metric) 133 mm
Width (Metric) 133 mm
Height (Metric) 128 mm