Learn More
Corning™ Polypropylene Conical Containers
Product Code. p-4429379
Click to view available options
Capacity (Metric):
200 mL
400 mL
1 L
Sterility:
Non-sterile
Sterile
Color:
Natural
White
Diameter (Metric):
62 mm
85 mm
105 mm
Unit Size:
Pack of 185
Pack of 220
Pack of 80
Product Code. 11838992
Brand: Corning™ PC200A02
Edge
Quality sampling tools for secure sample retrieval and shipment
  • Lightweight and break-resistant
  • Transparent or white Polypropylene (PP)
  • Assembled or non-assembled
TRUSTED_SUSTAINABILITY
Specifications
Graduated Yes
Material Polypropylene
Sterility Sterile
Capacity (English) 6.7 oz.
Capacity (Metric) 200 mL
Closure Color Natural
Closure Type Snap Cap
Color Natural
Diameter (Metric) 62 mm
Height (Metric) 88 mm
No. per Case 220/Cs.
No. per Pack 220/Pk.