missing translation for 'onlineSavingsMsg'
Learn More

Semadeni™ Polypropylene Containers

Brand:  Semadeni™ 2100

Additional Details : Weight : 0.01000kg

Product Code. 12637278

  • € 101.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Polypropylene
Chemical and Water Baths or as Storage
250L x 150W x 150 mmH
150 mm
Rectangle
Laboratory Container
250 mm
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers