Learn More
Polypropylene Containers
Brand: N/A D11390T
Additional Details : Weight : 4.50000kg
Specifications
125 mL | |
Transparent | |
Container | |
70 mm |
Brand: N/A D11390T
Additional Details : Weight : 4.50000kg
125 mL | |
Transparent | |
Container | |
70 mm |