Learn More
Polypropylene Containers
Click to view available options
Height (Metric):
68 mm
70 mm
Unit Size:
Pack of 1000
Pack of 300
Pack of 500
Specifications
Specifications
| Quantity | 500 Pk. |
| Capacity (Metric) | 125 mL |
| Color | Transparent |
| Product Type | Container |
| Height (Metric) | 68 mm |