Learn More
Buerkle™ Polypropylene Cylindrical 4-Way Hose Connectors
Product Code. 10723511 Shop All Bürkle Products
Click to view available options
Diameter (Metric):
11 to 13 mm
13 to 15 mm
3 to 5 mm
5 to 7 mm
7 to 9 mm
9 to 11 mm
Unit Size:
Pack of 10
Product Code. 10723511
Brand: Buerkle™ 87080010
Only null left
Add to basket
Add to basket
Polypropylene Cylindrical 4-Way Hose Connectors to connect hoses. Useful for extending, connecting, branching, joining and repairing hoses in the laboratory.
  • PP, Temperature resistant up to +135 °C max.
  • PVDF, Temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs, RoHS
Diameter (Metric) 9 to 11 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing with Diameter of 9 to 11 mm
Packaging Quantity 10/Pk.
Resistance Properties Up to 135°C Temperature-Resistant
Sterility Sterilizable
Shape Cylindrical X
Color Transparent
Max. Operating Temperature (Metric) 135°C
Product Type 4-Way Connector