Learn More
Polypropylene Disposable Tube
Product Code. 10536184
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 1000
Product Code. 10536184
Only null left
Specifications
Diameter (Metric) 16 mm
Product Type Disposable Tube
Closure Color White
Capacity (Metric) 10 mL
Material Polypropylene
Includes White screw cap
Packaging 1000Pack
Quantity 1000 Pack
Disposable Single-use
Closure Type Screw Cap