Learn More
Carl Roth™ Polypropylene Disposal Bags
Product Code. p-8006282
Encompass
Click to view available options
Thickness (Metric):
30 μm
Unit Size:
Pack of 500
Product Code. 10030672
Brand: Carl Roth™ H736.1
Only null left
Specifications
Disposable Single-use
For Use With (Application) Ideal for laboratory waste such as used pipettor tips, test tubes and vials.
Material Polypropylene
Thickness (Metric) 30 μm
Dimensions (L x W) 300 x 200mm
Length (Metric) 300 mm
Width (Metric) 200 mm
Product Type Waste Disposal Bag