Learn More
Cole-Parmer™ Polypropylene Dual-blade Paddle Assembly
Polypropylene Dual-blade Paddle Assembly for mixing of medium viscosity solutions.
Brand: Cole-Parmer™ 06367-10
Additional Details : Weight : 0.04000kg
Description
- 1 year warranty
Specifications
1.6 in. | |
41 mm | |
Use these plastic paddle assemblies for mixing most acids and corrosive solutions | |
350.83 mm | |
Dual-Blade Paddle Assembly |
0.3125 in. | |
8 mm | |
13.8 in. | |
1 Ea. | |
Polypropylene |