Learn More
Kappes Systeme™ Polypropylene Euro Transport Box
Marque: Kappes Systeme™ 6475.00.4150
Informations supplémentaires : Weight : 0.10000kg
Spécification
Transport Box | |
Blue | |
Conveyor, Grabber Technology, Storage and Retrieval Machine | |
1/Pk. | |
400 mm | |
180 mm |
Polypropylene | |
Rectangular | |
Food Safe | |
400 x 300 x 180 mm | |
300 mm |