Learn More
Kappes Systeme™ Polypropylene Euro Transport Box
Brand: Kappes Systeme™ 6475.00.4251
Additional Details : Weight : 0.10000kg
Specifications
Transport Box | |
Red | |
Conveyor, Grabber Technology, Storage and Retrieval Machine | |
1/Pk. | |
400 mm | |
220 mm |
Polypropylene | |
Rectangular | |
Food Safe | |
400 x 300 x 220 mm | |
300 mm |