Learn More
Kappes Systeme™ Polypropylene Euro Transport Box
Brand:  Kappes Systeme™ 6475.00.4051
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10768503
  • POA / Each
Estimated Shipment: 22-05-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Euro Transport Box
Red
1/Pk.
400 mm
120 mm
Polypropylene
Storage and retrieval machines, Compatible with standard european pallets
400 x 300 x 120 mm
300 mm