Learn More
Ark Plas Products™ Polypropylene Female Luer Lugs
Click to view available options
Port Size:
1/16 in. Barb
1/18 in. Barb
3/16 in. Barb
3/32 in. Barb
Unit Size:
Pack of 25
Description
- Natural polypropylene luer plugs
- Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
- Molded in virgin, lot-traceable material
- Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Specifications
| Certifications/Compliance | ISO 594-1 and 594-2 standards, RoHS Compliant, USP Class VI, FDA 21 CFR 177.1520, Animal Derivative-free, DEHP-free, BPA-free certified |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 4.8 mm |
| Material | Polypropylene |
| For Use With | Flexible tubing or threaded ports |
| Min. Operating Temperature (Metric) | -17°C |
| Packaging Quantity | 25/Pk. |
| Port Size | 3/16 in. Barb |
| Color | Natural |
| Max. Operating Temperature (Metric) | 135°C |
| Show More |