Learn More
BRAND™ Polypropylene Funnel with Fluted Interior
Product Code. 10140231 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric)Top:
65 mm
120 mm
150 mm
Diameter (Metric) Stem:
9 mm
11 mm
14 mm
Unit Size:
Each
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Diameter (Metric) Stem unitSize
10140231 65 mm 9 mm Pack of 20
10433711 120 mm 11 mm Each
10588901 150 mm 14 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10140231 Supplier BRAND™ Supplier No. 148220
Only null left
Add to basket
Add to basket
Polypropylene construction. Short wide stem provides smooth, quick funneling.
  • Polypropylene construction
  • Short, wide stem
  • Includes internal fluting
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 65 mm
Diameter (Metric) Stem 9 mm
Material Polypropylene
Length (Metric) Stem 63 mm