Learn More
Ratiolab™ Polypropylene Grid Inserts
Brand: Ratiolab™ 5220101
Additional Details : Weight : 0.19800kg
Specifications
Polypropylene | |
Insert Grid | |
5 Pack | |
133 mm |
Square | |
17.3 mm Diameter Tubes | |
133 mm | |
30 mm |