missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ VITsafe™ 500mL Wide-Mouth Wash Bottles

Product Code. 15732066
missing translation for 'orderingAttributeHoverText'
Chemical Label:
Acetone
Acetonitrile
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methanol
Methyl Ethyl Ketone
Methylene chloride
N,N-Dimethylformamide
Toluene
missing translation for 'unitSize'
Pack of 12
Pack of 6
This item is not returnable. View return policy

Product Code. 15732066

missing translation for 'mfr': VITLAB™ 1352949

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Imprinted with durable safety symbols, and other important information for optimum laboratory safety. Vitlab™ VITsafe™ Wide-Mouth Wash Bottles include color coded vent-cap that supports easy visual identification of the media.

  • Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
  • Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Imprinted with:
    • Material name in German, English, French and Spanish
    • Chemical formula, CAS No, hazard pictogram, signal word
    • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
  • The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping

Specifications

Capacity (Metric) 500 mL
Material LDPE
Closure Color Red
Certifications/Compliance Directive (EC) No. 1272/2008 (GHS)
Closure Included Yes
Mouth Wide
Height (Metric) 166 mm
Chemical Label Toluene
Closure Material Polypropylene
Diameter (Metric) 76 mm
Closure Type Vented
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.