Learn More
VITLAB™ Polypropylene Griffin Beakers
Click to view available options
Capacity (Metric):
10 mL
25 mL
5000 mL
Scale:
Blue Printed Scale
Raised and Embossed Blue Scale
Autoclavable:
Autoclavable
Conditionally
Unit Size:
Each
Pack of 12
Description
These low-form beakers with a convenient spout (Griffin beakers) are ideal for a variety of lab purposes and experiments. Lightweight polypropylene beakers with embossed graduations are convenient and easy to use.
- Constructed of durable, lightweight polypropylene that is heat resistant and chemically inert
- Highly transparent with a blue scale for easy identification, measuring and monitoring
- Printed graduations on smaller beakers (10 and 25mL)
- Raised, embossed graduations on 3,000 and 5,000mL beakers
- Ideal for a variety of lab purposes and experiments, including:
- Preparing solutions and decanting supernatant fluids
- Holding waste fluids prior to disposal
- Performing simple chemical reactions
- Manufactured according to ISO 7056 standards
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 38 mm |
| Capacity (Metric) | 25 mL |
| Material | Polypropylene |
| For Use With (Application) | For Volume Measurement |
| Height (Metric) | 50 mm |
| Increments | 5 mL |
| Quantity | 12 Pk. |
| Type | Beaker |
| Autoclavable | Conditionally |
| Show More |