Learn More
Brady™ Polypropylene: ISO Safety Sign - Wear safety harness
Product Code. 15684547
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
400 mm
Unit Size:
Each
Product Code. 15684547
Brand: Brady™ 820511
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M018-Dia 200-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
Diameter (Metric) 200 mm
For Use With (Application) Indoor and Outdoor
Thickness 1.501 mm
Temperature Range (Metric) -40°C to 120°C
Shape Round
Languages Global
Color White on Blue