Learn More
Mutexil™ Polypropylene Lab Coats
Product Code. p-8003232
Click to view available options
Size:
X-Large
2X-Large
Color:
Blue
White
Unit Size:
Pack of 50
Product Code. 11705423
Brand: Mutexil™ 4574713
Only null left
Add to basket
Edge
Add to basket
Polypropylene lab coats for use in laboratory settings. Available in a variety of sizes.
PP non woven labcoat with press-stud fastening and pockets
  • Disposable
  • PP, 40g/m2
  • For maintenance, production site visits
  • For non hazardous environments
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Polypropylene
Color Blue
Cuff Style Elastic
Disposable Single-use
Sterility Sterile
No. of Pockets 2
For Use With (Application) Non-hazardous environments with a high frequency change of clothing