Learn More
Masterflex™ Polypropylene Male Pipe Adapter Elbow Connector Barbed Fitting
Click to view available options
For Use With:
1/8 NPT Thread x 3/8 in. tubing I.D.
3/8 NPT Thread to 1/2 in. tubing I.D.
Unit Size:
Pack of 10
Specifications
Specifications
| Material | Polypropylene |
| For Use With | 1/8 NPT Thread x 3/8 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Port Size | 1/4 in. orifice; 1-13/32 x 1-13/32 in. |
| Thread Size | 1/8 in. NPT |
| Includes | NPT male pipe adapter elbow connector; ETO or gamma sterilization |
| Color | Natural |
| Product Type | Barbed Fitting |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering