Learn More
Cole-Parmer™ Polypropylene Manifolds
Product Code. 11755258
Click to view available options
Length (Metric):
79.7 mm
106.1 mm
155.9 mm
186.4 mm
238.3 mm
Unit Size:
Each
Product Code. 11755258
Brand: Cole-Parmer™ 3152225
Specifications
Type Manifold
Inlet 1/4 in.
Material Polypropylene
No. of Ports 10
Port Size (Metric) 4.3 mm
Length (Metric) 186.4 mm
Width (Metric) 25.4 mm
Height (Metric) 25.4 mm