Learn More
VITLAB™ Polypropylene Measuring Pipets
Brand:  VITLAB™ 163194
Additional Details : Weight : 1.23000kg
 View more versions of this product
Product Code. 15751986
  • € 86.65 / Pack of 12
Estimated Shipment
03-12-2020
Specifications
Yes
1 Pack
Measuring Pipette
Polypropylene
0.1 mL
±0.02 mL
300 mm
2 mL