missing translation for 'onlineSavingsMsg'
Learn More

ratiolab™ Makro Pipet Tips

Product Code. 10720553
Change view
Click to view available options
Volume (Metric):
1000 to 5000 μL
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10720553 1000 to 5000 μL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10720553 Supplier ratiolab™ Supplier No. 2400630
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these pipette tips are ideal for Gilson™ Pipetman™/Neo pipets. ratiolab™ Makro Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.

These pipette tips offer uniform high quality and precision. Together with specially developed Multirack, Refill System and the ratiolab™ Multibox with the associated pipet tips in Multitray, make pipetting much simpler and safe in the most sensitive applications
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types

Specifications

Volume (Metric) 1000 to 5000 μL
Color Translucent
Autoclavable Autoclavable
Length (Metric) 11.967cm
Certifications/Compliance CE, IVD
For Use With (Application) Ideal for All Commercially Available Pippets
Sterilization Method Autoclavable
For Use With (Equipment) Universal Fit
Product Line Makro
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.