missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Pipette Tips

Product Code. 12503267
Change view
Click to view available options
Volume (Metric):
0.1 to 10 mL
0.5 to 20 μL
1000 to 5000 μL
2 to 300 μL
Format:
Bulk
Rack
Refill
Color:
Light Blue
Natural
Neutral
Unit Size:
Pack of 1000
Pack of 200
Pack of 4800
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Format Color unitSize
12503267 0.5 to 20 μL Refill Neutral Pack of 4800
12533287 2 to 300 μL Bulk Neutral Pack of 1000
10358271 0.1 to 10 mL Rack Natural Pack of 960
10469371 1000 to 5000 μL Rack Light Blue Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12503267 Supplier Kartell™ Supplier No. 0974600
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.

  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
  • Disposable tips are also available racked

Specifications

Volume (Metric) 0.5 to 20 μL
Color Neutral
Autoclavable Autoclavable
Certifications/Compliance CE / IVD
Format Refill
For Use With (Equipment) Kartell Pluripet PL10, Eppendorf, Socorex, Hamilton
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.