Learn More
Karl Hecht™ Powder Funnel, Polypropylene
Product Code. 10328791
Click to view available options
Diameter (Metric)Top:
100mm
120mm
150mm
Unit Size:
Each
Product Code. 10328791
Brand: Karl Hecht™ 44002044
Only null left
Add to basket
Add to basket
Designed for the transfer of powdered materials. Karl Hecht™ Powder Funnel, Polypropylene facilitates pouring and reduces spilling risks.
  • Polypropylene construction.
  • Transparent.
  • Facilitates pouring operations.
  • Reduces the risk of spilling.
Specifications
Diameter (Metric)Top 120mm
Diameter (Metric) Stem 26mm
Material Polypropylene
For Use With (Application) Laboratory Applications
Length (Metric) Stem 20mm
Product Type Funnel