Learn More
Brady™ Polypropylene Prohibition Signs: Fire, Open Flame, and Smoking Prohibited
€ 13.25 - € 32.30
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10557614
|
Brady™
252003 |
200 x 200 mm |
€ 13.25
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10356194
|
Brady™
252004 |
315 x 315 mm |
€ 32.30
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||