missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Reagent Bottle
Store liquid and powder chemicals securely. VITLAB™ Polypropylene Reagent Bottle is constructed of transparent, durable polypropylene for long-term storage. Choose from neck-size stoppers or maximum grip screw caps, both available in a variety of sizes.
Brand: VITLAB™ 101594
This item is not returnable.
View return policy
Description
Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.- Transparent media bottles ideal for long-term storage of chemicals
- Reagent bottles featuring screw caps or neck stoppers
- Stoppers: neck-size with a hexagonal-knob cap and red core
- Screw caps: high-pitch GL threads for significant carrying power
- Larger sizes (5 and 10L) available with handles for easier carrying
- Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
- Constructed of study polypropylene
- Good chemical and temperature resistance
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction