Learn More
BRAND™ Polypropylene Reservoir
Reservoirs allow for different volumes and different solutions to be used in the automated liquid handling process.
Brand:  BRAND™ 701458
Additional Details : Weight : 0.31710kg
 View more versions of this product
Product Code. 15962957
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Pyramidal or flat bottom
  • DNase-free
  • RNase-free
Specifications
Specifications
Low Profile
Polypropylene
50 mL
1
50 mL/Column
Non-sterile
Yes
Flat