Learn More
Alpha Packaging™ Polypropylene Screw Caps
Use Alpha Packaging™ Polypropylene Screw Caps atop bottles containing analytical lab samples, especially those containing high purity chemicals, strong acids and solvents.
Brand: Alpha Packaging™ 307508
Additional Details : Weight : 0.00010kg
Description
- Autoclavable
- Polypropylene
- For use with screw-top bottles
Order Info
CertificationsCompliance
Specifications
Polypropylene | |
DIN 40 | |
DIN 40 | |
Polypropylene |