Learn More
Berlin Packaging™ Polypropylene Screw Caps
Product Code. 10136084
Click to view available options
Thread Size (Metric):
DIN 32
DIN 40
DIN 45
DIN 55
Unit Size:
Each
Pack of 14800
Pack of 26000
Pack of 60000
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10136084
Brand: Berlin Packaging™ 1001825
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Use Berlin Packaging™ Polypropylene Screw Caps atop bottles containing analytical lab samples, especially those containing high purity chemicals, strong acids and solvents.
  • Autoclavable
  • Polypropylene
  • For use with screw-top bottles
Specifications
Material Polypropylene
No. per Pack 1 Each
Quantity 1 Each
Thread Size (Metric) DIN 32
Neck Finish 07 mm
Product Type Screw Cap