Learn More
Berlin Packaging™ Polypropylene Screw Caps
Product Code. 10774927
Click to view available options
Thread Size (Metric):
DIN 32
DIN 40
DIN 45
DIN 55
Unit Size:
Each
Pack of 14800
Pack of 26000
Pack of 60000
Product Code. 10774927
Brand: Berlin Packaging™ 307509
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Berlin Packaging™ Polypropylene Screw Caps atop bottles containing analytical lab samples, especially those containing high purity chemicals, strong acids and solvents.
  • Autoclavable
  • Polypropylene
  • For use with screw-top bottles
Specifications
Material Polypropylene
No. per Pack 1 Each
Quantity 1 Each
Thread Size (Metric) DIN 45
Neck Finish 07 mm
Product Type Screw Cap