Learn More
Berlin Packaging™ Polypropylene Screw Caps
Click to view available options
Thread Size (Metric):
DIN 32
DIN 40
DIN 45
DIN 55
Unit Size:
Each
Pack of 14800
Pack of 26000
Pack of 60000
Description
- Autoclavable
- Polypropylene
- For use with screw-top bottles
Specifications
Specifications
| Material | Polypropylene |
| No. per Pack | 1 Each |
| Quantity | 1 Each |
| Thread Size (Metric) | DIN 55 |
| Neck Finish | 07 mm |
| Product Type | Screw Cap |