Learn More
Buerkle™ Polypropylene Measuring Scoop
Product Code. p-8011246 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
2 mL
5 mL
Length (Metric):
30 mm
82 mm
Width (Metric):
21 mm
27 mm
Unit Size:
Each
Product Code. 10780246
Brand: Buerkle™ 96140002
Only null left
Add to basket
Add to basket
Scoops and measures powders and granules. Buerkle™ Polypropylene Measuring Scoops are calibrated to accurately assess volume at the brim.
Universal graduated shovel for filling and measuring powders and granules. For exact metering, the graduated shovel is to be overfilled and the excess at the upper edge of the shovel is to be wiped off.
  • White polypropylene construction
  • High chemical-resistance
  • Calibrated volume when full to the brim
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2 mL
Color White
Product Type Measuring Scoop
Width (Metric) 21 mm
Dimensions (L x W) 30 x 21 mm
Material Polypropylene
Length (Metric) 30 mm